1-(4-Methoxyphenyl)tetrahydro-2H-imidazol-2-one

Lieferant: Apollo Scientific

Synonyme: 1-(4-Methoxyphenyl)imidazolidin-2-one

OR12110-1G OR12110-5G
APOSOR12110-1GEA 0 EUR
APOSOR12110-1G APOSOR12110-5G
1-(4-Methoxyphenyl)tetrahydro-2H-imidazol-2-one
1-(4-Methoxyphenyl)tetrahydro-2H-imidazol-2-one
Formel: C₁₀H₁₂N₂O₂
Molecular Weight: 192.22 g/mol
Schmelzpunkt: 210…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01720969
CAS-Nummer: 62868-39-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR