Methyl-2-acetyl-3-(dimethylamino)acrylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (E)-Methyl 2-((dimethylamino)methylene)-3-oxobutanoate, Methyl 2-[(dimethylamino)methylene]acetoacetate

OR12112-10G OR12112-1G OR12112-5G
APOSOR12112-10GEA 551 EUR
APOSOR12112-10G APOSOR12112-1G APOSOR12112-5G
Methyl-2-acetyl-3-(dimethylamino)acrylate 95%
Methyl-2-acetyl-3-(dimethylamino)acrylate
Formel: C₈H₁₃NO₃
Molecular Weight: 171.2 g/mol
Schmelzpunkt: 66…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689663
CAS-Nummer: 203186-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR