6-Methyl-1H-indazol-5-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-6-methyl-1H-indazole

OR12141-1G OR12141-250MG
APOSOR12141-1GEA 111 EUR
APOSOR12141-1G APOSOR12141-250MG
6-Methyl-1H-indazol-5-amine
6-Methyl-1H-indazol-5-amine
Formel: C₈H₉N₃
Molecular Weight: 147.18 g/mol
Schmelzpunkt: 215…217 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD03305372
CAS-Nummer: 81115-45-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR