1-Benzyl-2-[4-(tert-butyl)phenyl]-6-oxo-3-piperidinecarboxylic acid

Lieferant: Apollo Scientific

OR12169-5G OR12169-10G OR12169-1G
APOSOR12169-5GEA 538 EUR
APOSOR12169-5G APOSOR12169-10G APOSOR12169-1G
1-Benzyl-2-[4-(tert-butyl)phenyl]-6-oxo-3-piperidinecarboxylic acid
1-Benzyl-2-[4-(tert-butyl)phenyl]-6-oxo-3-piperidinecarboxylic acid
Formel: C₂₃H₂₇NO₃
Molecular Weight: 365,48 g/mol
Schmelzpunkt: 144…146 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027174
CAS-Nummer: 952182-64-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR