4,6-Dichlornicotinsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,6-dichloropyridine-3-carboxylic acid

OR12186-1G OR12186-5G OR12186-10G
APOSOR12186-1GEA 21.9 EUR
APOSOR12186-1G APOSOR12186-5G APOSOR12186-10G
4,6-Dichlornicotinsäure 98%
4,6-Dichloronicotinic acid
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192,00 g/mol
Schmelzpunkt: 152…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234146
CAS-Nummer: 73027-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR