[(1-Methyl-1H-pyrazol-3-yl)methyl]amine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(1-Methyl-1H-pyrazol-3-yl)methanamine
OR12222-1G
OR12222-250MG
APOSOR12222-1GEA
102
EUR
APOSOR12222-1G
APOSOR12222-250MG
[(1-Methyl-1H-pyrazol-3-yl)methyl]amine
[(1-Methyl-1H-pyrazol-3-yl)methyl]amine
Formel:
C₅H₉N₃ Molecular Weight: 111.15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419807 CAS-Nummer: 612511-81-6 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...