[4-(Aminomethyl)tetrahydro-2H-pyran-4-yl]dimethylamine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Aminomethyl)-4-(dimethylamino)tetrahydro-2H-pyran

OR12234-1G OR12234-250MG
APOSOR12234-1GEA 531 EUR
APOSOR12234-1G APOSOR12234-250MG
[4-(Aminomethyl)tetrahydro-2H-pyran-4-yl]dimethylamine 95%
[4-(Aminomethyl)tetrahydro-2H-pyran-4-yl]dimethylamine
Formel: C₈H₁₈N₂O
Molecular Weight: 158,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09065027
CAS-Nummer: 176445-80-0
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR