3-Isochinolinol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Hydroxyisoquinoline
OR1224-5G
OR1224-100G
OR1224-25G
APOSOR1224-5GEA
139
EUR
APOSOR1224-5G
APOSOR1224-100G
APOSOR1224-25G
3-Isochinolinol
3-Isochinolinol
Formel:
C₉H₇NO Molecular Weight: 145.16 g/mol Schmelzpunkt: 195…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075524 CAS-Nummer: 7651-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...