7-Benzo[b]thiophencarbonsäure 97%

Lieferant: Apollo Scientific
Achtung

OR12294-1G OR12294-250MG
APOSOR12294-1GEA 285 EUR
APOSOR12294-1G APOSOR12294-250MG
7-Benzo[b]thiophencarbonsäure 97%
7-Benzo[b]thiophencarbonsäure
Formel: C₉H₆O₂S
Schmelzpunkt: 176,5…178.5 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 10134-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR