1-Azepanyl(3-piperidinyl)methanone 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Piperidin-3-ylcarbonyl)azepane
OR12324-1G
OR12324-5G
APOSOR12324-1GEA
94.4
EUR
APOSOR12324-1G
APOSOR12324-5G
1-Azepanyl(3-piperidinyl)methanone 97%
1-Azepanyl(3-piperidinyl)methanone
Formel:
C₁₂H₂₂N₂O Molecular Weight: 210,32 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06016189 CAS-Nummer: 690632-28-1 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...