tert-Butyl-4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-(4-cyanophenyl)piperazine-1-carboxylate, 4-(Piperazin-4-yl)benzonitrile, N1-BOC protected
OR12333-1G
OR12333-5G
APOSOR12333-1GEA
21.9
EUR
APOSOR12333-1G
APOSOR12333-5G
tert-Butyl-4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate 97%
tert-Butyl-4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Formel:
C₁₆H₂₁N₃O₂ Molecular Weight: 287,36 g/mol Schmelzpunkt: 123…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791198 CAS-Nummer: 186650-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...