dibenzo[b,d]furan-2-carbaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
dibenzo[b,d]furan-2-carbaldehyde, Dibenzofuran-2-carboxaldehyde
OR12348-1G
OR12348-250MG
OR12348-5G
APOSOR12348-1GEA
48
EUR
APOSOR12348-1G
APOSOR12348-250MG
APOSOR12348-5G
dibenzo[b,d]furan-2-carbaldehyde 97%
dibenzo[b,d]furan-2-carbaldehyde
Formel:
C₁₃H₈O₂ Molecular Weight: 196,205 g/mol Schmelzpunkt: 73…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00136272 CAS-Nummer: 5397-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...