2-Bromo-3-furoyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR12387-100MG
OR12387-250MG
OR12387-500MG
APOSOR12387-100MGEA
144
EUR
APOSOR12387-100MG
APOSOR12387-250MG
APOSOR12387-500MG
2-Bromo-3-furoyl chloride
2-Bromo-3-furoyl chloride
Formel:
C₅H₂BrClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09702418 CAS-Nummer: 915707-69-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...