(1-Methyl-1H-pyrazol-5-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Hydroxymethyl)-1-methylpyrazole, 1-Methylpyrazole-5-methanol
OR12407-1G
OR12407-250MG
OR12407-5G
APOSOR12407-1GEA
58
EUR
APOSOR12407-1G
APOSOR12407-250MG
APOSOR12407-5G
(1-Methyl-1H-pyrazol-5-yl)methanol
(1-Methyl-1H-pyrazol-5-yl)methanol
Formel:
C₅H₈N₂O Molecular Weight: 112,13 g/mol Schmelzpunkt: 57…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08556301 CAS-Nummer: 84547-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...