Methyl-3-amino-5-hydroxybenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-amino-5-hydroxybenzoate
OR12411-1G
OR12411-25G
OR12411-5G
APOSOR12411-1GEA
174
EUR
APOSOR12411-1G
APOSOR12411-25G
APOSOR12411-5G
Methyl-3-amino-5-hydroxybenzoat
Methyl-3-amino-5-hydroxybenzoat
Formel:
C₈H₉NO₃ Molecular Weight: 167,1620 g/mol Schmelzpunkt: 124…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09263220 CAS-Nummer: 67973-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...