4-(Brommethyl)-3-phenylisoxazol
Lieferant: Apollo Scientific
|
Gefahr
|
OR12434-1G
OR12434-500MG
APOSOR12434-1GEA
0
EUR
APOSOR12434-1G
APOSOR12434-500MG
4-(Brommethyl)-3-phenylisoxazol
4-(Bromomethyl)-3-phenylisoxazole
Formel:
C₁₀H₈BrNO Schmelzpunkt: 82,5…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159724 CAS-Nummer: 2039-50-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...