tert-Butyl-4-(3-ethoxy-3-oxopropanoyl)tetrahydro-1(2H)-pyridinecarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Boc-4-(2-Ethoxycarbonyl-acetyl)piperidine

OR12465-5G
APOSOR12465-5GEA 55.1 EUR
APOSOR12465-5G
tert-Butyl-4-(3-ethoxy-3-oxopropanoyl)tetrahydro-1(2H)-pyridinecarboxylate
tert-Butyl-4-(3-ethoxy-3-oxopropanoyl)tetrahydro-1(2H)-pyridinecarboxylate
Formel: C₁₅H₂₅NO₅
Molecular Weight: 299,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115569
CAS-Nummer: 479630-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR