Methyl amino(3,4-dichlorophenyl)acetate hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
3,4-Dichloro-DL-phenylglycine methyl ester hydrochloride
OR12471-1G
OR12471-250MG
APOSOR12471-1GEA
452
EUR
APOSOR12471-1G
APOSOR12471-250MG
Methyl amino(3,4-dichlorophenyl)acetate hydrochloride 95%
Methyl amino(3,4-dichlorophenyl)acetate hydrochloride
Formel:
C₉H₁₀Cl₃NO₂ Molecular Weight: 270,54 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115512 CAS-Nummer: 1078611-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...