2-Phenylpyrrolidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR12503-5G
APOSOR12503-5GEA
58
EUR
APOSOR12503-5G
2-Phenylpyrrolidine 97%
2-Phenylpyrrolidine
Formel:
C₁₀H₁₃N Molecular Weight: 147.2200 g/mol Siedepunkt: 114 °C/7.5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631835 CAS-Nummer: 1006-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...