3-Phenylpyrrolidine
Lieferant: Apollo Scientific
|
Achtung
|
OR12549-1G
OR12549-250MG
OR12549-100MG
APOSOR12549-1GEA
50.9
EUR
APOSOR12549-1G
APOSOR12549-250MG
APOSOR12549-100MG
3-Phenylpyrrolidine
3-Phenylpyrrolidine
Formel:
C10H13N Molecular Weight: 147,22 g/mol Siedepunkt: 92…94 °C/2mmHg Dichte: 1,027 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD01838169 CAS-Nummer: 936-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...