Methyl-3-methyl-2-furoat 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-methylfuran-2-carboxylate

OR12570-5G
APOSOR12570-5GEA 77 EUR
APOSOR12570-5G
Methyl-3-methyl-2-furoat 98%
Methyl-3-methyl-2-furoat
Formel: C₇H₈O₃
Molecular Weight: 140.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014110
CAS-Nummer: 6141-57-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR