3-(2-Pyridyl)benzaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Pyridin-2-yl)benzaldehyde

OR12577-1G OR12577-250MG OR12577-5G
APOSOR12577-1GEA 56.7 EUR
APOSOR12577-1G APOSOR12577-250MG APOSOR12577-5G
3-(2-Pyridyl)benzaldehyd
3-(2-Pyridyl)benzaldehyd
Formel: C₁₂H₉NO
Molecular Weight: 183,2060 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684103
CAS-Nummer: 85553-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR