4-(3-Pyridyl)benzaldehyd 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Pyridin-3-yl)benzaldehyde

OR12649-1G OR12649-25G OR12649-5G
APOSOR12649-1GEA 68.3 EUR
APOSOR12649-1G APOSOR12649-25G APOSOR12649-5G
4-(3-Pyridyl)benzaldehyd 95%
4-(3-Pyridyl)benzaldehyd
Formel: C₁₂H₉NO
Molecular Weight: 183,2060 g/mol
Schmelzpunkt: 51,7…53.7 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631910
CAS-Nummer: 127406-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR