Ethyl-5-(2-bromoacetyl)isoxazole-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR1266-250MG
OR1266-1G
APOSOR1266-250MGEA
46.4
EUR
APOSOR1266-250MG
APOSOR1266-1G
Ethyl-5-(2-bromoacetyl)isoxazole-3-carboxylate
Ethyl-5-(2-bromoacetyl)isoxazole-3-carboxylate
Formel:
C₈H₈BrNO₄ Molecular Weight: 262,07 g/mol Schmelzpunkt: 71…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173773 CAS-Nummer: 104776-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...