Methyl-3-(4-(bromomethyl)phenyl)acrylate 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-(4-(bromomethyl)phenyl)acrylate, Methyl 4-(bromomethyl)cinnamate
OR12661-100G
APOSOR12661-100GEA
150
EUR
APOSOR12661-100G
Methyl-3-(4-(bromomethyl)phenyl)acrylate 96%
Methyl-3-(4-(bromomethyl)phenyl)acrylate
Formel:
C₁₁H₁₁BrO₂ Molecular Weight: 255,1100 g/mol Schmelzpunkt: 58…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02684186 CAS-Nummer: 946-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...