2-(Methylsulfonyl)ethanaminhydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Methylsulphonyl)ethanamine hydrochloride, 2-Aminoethyl Methyl sulphone Hydrochloride, 2-Aminoethyl methyll sulphone hydrochloride, 2-(Methylsulphonyl)ethanamine Hydrochloride
OR12674-500MG
OR12674-1G
APOSOR12674-500MGEA
21.9
EUR
APOSOR12674-500MG
APOSOR12674-1G
2-(Methylsulfonyl)ethanaminhydrochlorid
2-(Methylsulphonyl)ethanamine hydrochloride
Formel:
C₃H₁₀ClNO₂S Molecular Weight: 159,6400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840162 CAS-Nummer: 104458-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...