2-Methyl-1,3-benzothiazol-6-amine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Amino-2-methyl-1,3-benzothiazole
OR12777-1G
APOSOR12777-1GEA
65.4
EUR
APOSOR12777-1G
2-Methyl-1,3-benzothiazol-6-amine 98%
2-Methyl-1,3-benzothiazol-6-amine
Formel:
C₈H₈N₂S Molecular Weight: 164,23 g/mol Schmelzpunkt: 121…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00160076 CAS-Nummer: 2941-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...