5-Bromindol 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-1H-indole
OR1278-100G
OR1278-500G
APOSOR1278-100GEA
31.9
EUR
APOSOR1278-100G
APOSOR1278-500G
5-Bromindol 99%
5-Bromindol
Formel:
C₈H₆BrN Molecular Weight: 196,05 g/mol Schmelzpunkt: 90…92 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005670 CAS-Nummer: 10075-50-0 EINECS: 233-208-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...