4-(4-Chlorphenoxy)piperidin Hydrochlorid 98%

Lieferant: Apollo Scientific
Achtung

OR12798-1G
APOSOR12798-1GEA 190 EUR
APOSOR12798-1G
4-(4-Chlorphenoxy)piperidin Hydrochlorid 98%
4-(4-Chlorphenoxy)piperidin Hydrochlorid
Formel: C₁₁H₁₅Cl₂NO
Schmelzpunkt: 194…196 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 63843-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR