1-Phenyl-1,3-butandion
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzoylaceton, 1-Phenylbutan-1,3-dion
OR12814-5G
OR12814-25G
OR12814-100G
APOSOR12814-5GEA
21.9
EUR
APOSOR12814-5G
APOSOR12814-25G
APOSOR12814-100G
1-Phenyl-1,3-butandion
1-Phenyl-1,3-butandion
Formel:
C₆H₅C(O)CH₂C(O)CH₃ Molecular Weight: 162,19 g/mol Schmelzpunkt: 54…56 °C Dichte: 1,09 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008786 CAS-Nummer: 93-91-4 EINECS: 202-286-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...