3,5-Dimethyl-4-isoxazolcarbonylchlorid 95%

Lieferant: Apollo Scientific
Gefahr

OR1284-5G OR1284-1G
APOSOR1284-5GEA 145 EUR
APOSOR1284-5G APOSOR1284-1G
3,5-Dimethyl-4-isoxazolcarbonylchlorid 95%
3,5-Dimethyl-4-isoxazolcarbonylchlorid
Formel: C₆H₆ClNO₂
Molecular Weight: 159.57 g/mol
Siedepunkt: 90 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00052555
CAS-Nummer: 31301-45-8
UN: 1760
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR