4,4-Diphenylpiperidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR12865-1G
OR12865-250MG
OR12865-5G
APOSOR12865-1GEA
244
EUR
APOSOR12865-1G
APOSOR12865-250MG
APOSOR12865-5G
4,4-Diphenylpiperidine hydrochloride
4,4-Diphenylpiperidine hydrochloride
Formel:
C₁₇H₂₀ClN Lagertemperatur: Raumtemperatur |
MDL:
MFCD01700799 CAS-Nummer: 63675-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...