Methyl-4-oxo-3-piperidincarboxylathydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-oxopiperidine-3-carboxylate hydrochloride, Methyl 4-Oxopiperidine-3-carboxylate HCl
OR12915-5G
OR12915-25G
APOSOR12915-5GEA
21.9
EUR
APOSOR12915-5G
APOSOR12915-25G
Methyl-4-oxo-3-piperidincarboxylathydrochlorid
Methyl-4-oxopiperidine-3-carboxylate hydrochloride
Formel:
C₇H₁₂ClNO₃ Molecular Weight: 193,63 g/mol Schmelzpunkt: 173 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012769 CAS-Nummer: 71486-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...