4-Nitrobenzolsulfonsäurehydrat
Lieferant: Apollo Scientific
|
Gefahr
|
OR12921-100G
OR12921-25G
OR12921-5G
APOSOR12921-100GEA
319
EUR
APOSOR12921-100G
APOSOR12921-25G
APOSOR12921-5G
4-Nitrobenzolsulfonsäurehydrat
4-Nitrobenzolsulfonsäurehydrat
Formel:
C₆H₅NO₅S Molecular Weight: 203,17 g/mol Schmelzpunkt: 95…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041886 CAS-Nummer: 138-42-1 EINECS: 205-329-5 UN: 2305 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...