1,2,3-Thiadiazol-4-carbonsäure

Lieferant: Apollo Scientific
Achtung

OR1294-5G
APOSOR1294-5GEA 172 EUR
APOSOR1294-5G
1,2,3-Thiadiazol-4-carbonsäure
1,2,3-Thiadiazole-4-carboxylic acid
Formel: C₃H₂N₂O₂S
Schmelzpunkt: 227…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052099
CAS-Nummer: 4100-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR