Ethyl-2-amino-4-(2-thienyl)thiophene-3-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5'-amino-2,3'-bithiophene-4'-carboxylate

OR1296-10G OR1296-1G OR1296-5G
APOSOR1296-10GEA 519 EUR
APOSOR1296-10G APOSOR1296-1G APOSOR1296-5G
Ethyl-2-amino-4-(2-thienyl)thiophene-3-carboxylate
Ethyl-2-amino-4-(2-thienyl)thiophene-3-carboxylate
Formel: C₁₁H₁₁NO₂S₂
Molecular Weight: 253.34 g/mol
Schmelzpunkt: 84…87 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00115120
CAS-Nummer: 243669-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR