4-Pyridin-3-ylpiperidin-4-ol dihydrochloride

Lieferant: Apollo Scientific
Achtung

OR12982-1G OR12982-250MG
APOSOR12982-1GEA 30.6 EUR
APOSOR12982-1G APOSOR12982-250MG
4-Pyridin-3-ylpiperidin-4-ol dihydrochloride
4-Pyridin-3-ylpiperidin-4-ol dihydrochloride
Formel: C₁₀H₁₆Cl₂N₂O
Molecular Weight: 251.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16988629
CAS-Nummer: 50461-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR