Ethyl-2-methylthiazol-4-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-methylthiazole-4-carboxylate
OR1301-10G
OR1301-1G
OR1301-5G
APOSOR1301-10GEA
34.8
EUR
APOSOR1301-10G
APOSOR1301-1G
APOSOR1301-5G
Ethyl-2-methylthiazol-4-carboxylat
Ethyl-2-methylthiazol-4-carboxylat
Formel:
C₇H₉NO₂S Molecular Weight: 171.22 g/mol Schmelzpunkt: 54…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156148 CAS-Nummer: 6436-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...