(3,5-Dimethoxy-4-hydroxyphenyl)essigsäure 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Hydroxy-3,5-dimethoxyphenyl)acetic acid, 4-Hydroxy-3,5-dimethoxyphenylacetic acid

OR13021-1G OR13021-25G OR13021-5G
APOSOR13021-1GEA 33.5 EUR
APOSOR13021-1G APOSOR13021-25G APOSOR13021-5G
(3,5-Dimethoxy-4-hydroxyphenyl)essigsäure 99%
(3,5-Dimethoxy-4-hydroxyphenyl)essigsäure
Formel: C₁₀H₁₂O₅
Molecular Weight: 212,2 g/mol
Schmelzpunkt: 127…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016834
CAS-Nummer: 4385-56-2
EINECS: 224-494-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR