4-Brom-1-naphthylamin

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Amino-4-bromnaphthalin

OR13038-100G OR13038-25G OR13038-5G
APOSOR13038-100GEA 106 EUR
APOSOR13038-100G APOSOR13038-25G APOSOR13038-5G
4-Brom-1-naphthylamin
4-Brom-1-naphthylamin
Formel: C₁₀H₈BrN
Molecular Weight: 222.08 g/mol
Schmelzpunkt: 102…103 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00004023
CAS-Nummer: 2298-07-9
EINECS: 218-944-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR