4-Aminophenylboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester

OR13050-1G OR13050-25G OR13050-5G
APOSOR13050-1GEA 21.9 EUR
APOSOR13050-1G APOSOR13050-25G APOSOR13050-5G
4-Aminophenylboronsäurepinakolester
4-Aminophenylboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
Molecular Weight: 219,09 g/mol
Schmelzpunkt: 165…171 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02093721
CAS-Nummer: 214360-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR