4-Hydroxy-6-methylcoumarin

Lieferant: Apollo Scientific
Achtung

OR13053-1G OR13053-5G
APOSOR13053-1GEA 59.6 EUR
APOSOR13053-1G APOSOR13053-5G
4-Hydroxy-6-methylcoumarin
4-Hydroxy-6-methylcoumarin
Formel: C₁₀H₈O₃
Molecular Weight: 176,17 g/mol
Schmelzpunkt: 261…264 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00085738
CAS-Nummer: 13252-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR