1-Benzyloxy-3-brombenzol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Benzyloxybromobenzene, 1-(Benzyloxy)-3-bromobenzene
OR13056-25G
OR13056-5G
OR13056-100G
APOSOR13056-25GEA
127
EUR
APOSOR13056-25G
APOSOR13056-5G
APOSOR13056-100G
1-Benzyloxy-3-brombenzol
1-Benzyloxy-3-brombenzol
Formel:
C₁₃H₁₁BrO Molecular Weight: 263.13 Schmelzpunkt: 57…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155065 CAS-Nummer: 53087-13-1 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...