4-Amino-2-brompyridin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Brom-4-pyridylamin, 2-Brompyridin-4-amin
OR13064-25G
OR13064-5G
OR13064-100G
APOSOR13064-25GEA
89.9
EUR
APOSOR13064-25G
APOSOR13064-5G
APOSOR13064-100G
4-Amino-2-brompyridin
4-Amino-2-brompyridin
Formel:
C₅H₅BrN₂ Molecular Weight: 173,01 g/mol Schmelzpunkt: 92…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646061 CAS-Nummer: 7598-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...