3-(Chlormethyl)-5-methylisoxazol

Lieferant: Apollo Scientific
Gefahr

OR1308-1G OR1308-5G OR1308-250MG
APOSOR1308-1GEA 69.6 EUR
APOSOR1308-1G APOSOR1308-5G APOSOR1308-250MG
3-(Chlormethyl)-5-methylisoxazol
3-(Chloromethyl)-5-methylisoxazole
Formel: C₅H₆ClNO
Molecular Weight: 131,560 g/mol
Siedepunkt: 93 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00221166
CAS-Nummer: 35166-37-1
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR