Bis(4-bromophenyl)acetylene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Bis(4-bromophenyl)ethyne
OR13096-1G
OR13096-25G
OR13096-5G
APOSOR13096-1GEA
21.9
EUR
APOSOR13096-1G
APOSOR13096-25G
APOSOR13096-5G
Bis(4-bromophenyl)acetylene
Bis(4-bromophenyl)acetylene
Formel:
C₁₄H₈Br₂ Molecular Weight: 336,03 g/mol Schmelzpunkt: 183…187 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00185147 CAS-Nummer: 2789-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...