4-Chlor-3-nitrophenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-3-nitrobenzeneboronic acid

OR13107-1G OR13107-5G OR13107-25G
APOSOR13107-1GEA 21.9 EUR
APOSOR13107-1G APOSOR13107-5G APOSOR13107-25G
4-Chlor-3-nitrophenylboronsäure
4-Chlor-3-nitrophenylboronsäure
Formel: C₆H₅BClNO₄
Molecular Weight: 201.37 g/mol
Schmelzpunkt: 275 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02258950
CAS-Nummer: 151169-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR