(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-(hydroxymethyl)benzeneboronic acid dehydrate hydrochloride, 6-Aminobenzo[c][1,2]oxaborol-1(3H)-ol hydrochloride

OR13108-1G OR13108-250MG
APOSOR13108-1GEA 200 EUR
APOSOR13108-1G APOSOR13108-250MG
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride
Formel: C₇H₉BClNO₂
Molecular Weight: 185,42 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115645
CAS-Nummer: 117098-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR