1-Cyclopentenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Cyclopent-1-en-1-ylboronic acid
OR13114-1G
OR13114-5G
OR13114-25G
OR13114-100G
APOSOR13114-1GEA
21.9
EUR
APOSOR13114-1G
APOSOR13114-5G
APOSOR13114-25G
APOSOR13114-100G
1-Cyclopentenylboronsäure 97%
1-Cyclopentenylboronsäure
Formel:
C₅H₉BO₂ Molecular Weight: 111,94 g/mol Schmelzpunkt: 134…140 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179496 CAS-Nummer: 850036-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...