2-Hydroxymethyl-5-nitrobenzeneboronic acid hemiester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Nitrobenzo[c][1,2]oxaborol-1(3H)-ol, 2-(Hydroxymethyl)-5-nitrobenzeneboronic acid dehydrate
OR13117-250MG
OR13117-1G
OR13117-5G
APOSOR13117-250MGEA
116
EUR
APOSOR13117-250MG
APOSOR13117-1G
APOSOR13117-5G
2-Hydroxymethyl-5-nitrobenzeneboronic acid hemiester
2-Hydroxymethyl-5-nitrobenzeneboronic acid hemiester
Formel:
C₇H₆BNO₄ Molecular Weight: 178,94 g/mol Schmelzpunkt: 176…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115657 CAS-Nummer: 118803-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...